Learn More
Medchemexpress LLC Benzoic acid, 2-amino-6-methyl- | 4389-50-8 | 99.4% | 151.16 | 10 G
SDP
Catalog No. 5000404511
Encompass_Preferred
2-Amino-6-methylbenzoic acid is a biochemical reagent suitable for life science-related research, where it can be utilized as a biological material or an organic compound. It is specifically mentioned that this compound reacts with cyanogen bromide to form 2-amino-5-methyl-benzo[d]oxazin-4-one. The product is intended for research use only.
  • Biochemical reagent for life science research
  • Can be used as a biological material
  • Can be used as an organic compound
  • High purity: 99.36%
Catalog No. 50-004-04511 Supplier Medchemexpress LLC Supplier No. HY4149610G
2-Amino-6-methylbenzoic acid is a biochemical reagent suitable for life science-related research, where it can be utilized as a biological material or an organic compound. It is specifically mentioned that this compound reacts with cyanogen bromide to form 2-amino-5-methyl-benzo[d]oxazin-4-one. The product is intended for research use only.
  • Biochemical reagent for life science research
  • Can be used as a biological material
  • Can be used as an organic compound
  • High purity: 99.36%