Learn More
Medchemexpress LLC Benzoic acid, 2-formyl- | 119-67-5 | MFCD00003336 | 100g
SDP
Supplier:  Medchemexpress LLC HYY1771100G
Encompass_Preferred
2-Carboxybenzaldehyde is a key intermediate metabolite in the biodegradation of polycyclic aromatic hydrocarbons (PAHs) such as fluoranthene and phenanthrene 2-Carboxybenzaldehyde can be produced by the degradation of fluoranthene by Pasteurella sp IFA and Mycobacterium sp PYR-1 2-Carboxybenzaldehyde can also be produced by the degradation of phenanthrene by Pseudomonas sp PPD[1][2][3]
Catalog No. 50-002-66250