Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 3-(1,3,3a,4,9,9a-hexahydro-4-nitro-1,3-dioxo-4,9[1',2']-benzeno-2H-isoindol-2-yl) | 690991-06-1 | 99.7% | 440.40 g·mol⁻1 | C25H16N2O6 | 5 MG
SDP

Catalog No. 5000223088
Encompass_Preferred

S100P-IN-1 is a small-molecule inhibitor of S100P that blocks S100P-RAGE binding and reduces invasive behavior in S100P-expressing pancreatic cancer cells. It is intended for in vitro research of S100P-related signaling and invasion assays.

  • Potent S100P inhibition with reported IC50 22.7 nM.
  • Anti-metastatic activity in pancreatic cancer cell invasion assays at 10 μM without obvious cytotoxicity.
  • Molecular formula C25H16N2O6 and molecular weight 440.40 g·mol⁻1.
  • High purity (99.74%).
  • Available as solid and as 10 mM solution in DMSO in multiple package sizes.
  • Storage: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • Suitable for in vitro studies of S100P-related pathways and invasion assays.

Catalog No. 50-002-23088 Supplier Medchemexpress LLC Supplier No. HY1466955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S100P-IN-1 is a small-molecule inhibitor of S100P that blocks S100P-RAGE binding and reduces invasive behavior in S100P-expressing pancreatic cancer cells. It is intended for in vitro research of S100P-related signaling and invasion assays.

  • Potent S100P inhibition with reported IC50 22.7 nM.
  • Anti-metastatic activity in pancreatic cancer cell invasion assays at 10 μM without obvious cytotoxicity.
  • Molecular formula C25H16N2O6 and molecular weight 440.40 g·mol⁻1.
  • High purity (99.74%).
  • Available as solid and as 10 mM solution in DMSO in multiple package sizes.
  • Storage: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month).
  • Suitable for in vitro studies of S100P-related pathways and invasion assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.