Learn More
Medchemexpress LLC Benzoic acid, 3-[[[2-[4-[2-[[4-[[3-(aminocarbonyl)phenyl]amino]-1,4-dioxobutyl]amino]ethyl]-1H-1,2,3-triazol-1-yl]ethyl]amino]sulfonyl]- | 2084811-68-5 | MFCD32215309 | 99.5% | 557.58 | C24H27N7O7S | 1 MG
SDP
Catalog No. 5000228307
Encompass_Preferred
PARP14 inhibitor H10 is a selective small-molecule research reagent that targets PARP14. It inhibits PARP14 with an IC50 of 490 nM, shows approximately 24-fold selectivity over PARP1, and has been reported to induce caspase-3/7-mediated apoptosis in cellular studies. Supplied as a solid for laboratory research use only.
  • Selective PARP14 inhibition (IC50 = 490 nM).
  • Approximately 24-fold selectivity over PARP1.
  • Reported to induce caspase-3/7-mediated apoptosis in cells.
  • High reported purity suitable for research applications.
  • Provided in small milligram pack sizes for experimental workflows.
Catalog No. 50-002-28307 Supplier Medchemexpress LLC Supplier No. HY1178891MG
PARP14 inhibitor H10 is a selective small-molecule research reagent that targets PARP14. It inhibits PARP14 with an IC50 of 490 nM, shows approximately 24-fold selectivity over PARP1, and has been reported to induce caspase-3/7-mediated apoptosis in cellular studies. Supplied as a solid for laboratory research use only.
  • Selective PARP14 inhibition (IC50 = 490 nM).
  • Approximately 24-fold selectivity over PARP1.
  • Reported to induce caspase-3/7-mediated apoptosis in cells.
  • High reported purity suitable for research applications.
  • Provided in small milligram pack sizes for experimental workflows.