Learn More
Medchemexpress LLC Benzoic acid, 3-[[[3-[4-[(methylsulfonyl)amino]phenyl]-4-oxo-4H-1-benzopyran-7-yl]oxy]methyl]. | 1005334-57-5 | 99.9% | 10 MG
SDP
Catalog No. 5000297991
Encompass_Preferred
CVT-10216 is a small-molecule, selective reversible inhibitor of aldehyde dehydrogenase-2 supplied for research use. It is provided as a high-purity powder for preclinical and biochemical studies investigating alcohol consumption, anxiolytic effects, and aldehyde metabolism, with supporting analytical documentation and storage recommendations.
  • Selective reversible inhibition of ALDH-2 with low nanomolar potency.
  • High reported purity suitable for analytical and in vivo studies.
  • Provided as a powder in mg-scale pack sizes for laboratory use.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Recommended storage conditions specified for powder and solutions.
Catalog No. 50-002-97991 Supplier Medchemexpress LLC Supplier No. HY1980110MG
CVT-10216 is a small-molecule, selective reversible inhibitor of aldehyde dehydrogenase-2 supplied for research use. It is provided as a high-purity powder for preclinical and biochemical studies investigating alcohol consumption, anxiolytic effects, and aldehyde metabolism, with supporting analytical documentation and storage recommendations.
  • Selective reversible inhibition of ALDH-2 with low nanomolar potency.
  • High reported purity suitable for analytical and in vivo studies.
  • Provided as a powder in mg-scale pack sizes for laboratory use.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Recommended storage conditions specified for powder and solutions.