Learn More
Medchemexpress LLC Benzoic acid, 3,4,5-trihydroxy-, hydrate (1:1) | 5995-86-8 | 188.13 | 25 MG
SDP
Catalog No. 5000390254
Encompass_Preferred
Gallic acid hydrate (3,4,5-Trihydroxybenzoic acid hydrate) is a natural polyhydroxyphenolic compound and a free radical scavenger. It inhibits cyclooxygenase-2 (COX-2) and exhibits a range of biological activities including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties. This product is intended for research use only.
  • Natural polyhydroxyphenolic compound
  • Acts as a free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial activity
  • Provides antioxidant properties
  • Demonstrates anti-inflammatory effects
  • Possesses anticancer activities
  • Improves glucose tolerance in vivo
  • Reduces serum triglyceride concentration in vivo
  • Intended for research use only
Catalog No. 50-003-90254 Supplier Medchemexpress LLC Supplier No. 25MGHYN0523AR
Gallic acid hydrate (3,4,5-Trihydroxybenzoic acid hydrate) is a natural polyhydroxyphenolic compound and a free radical scavenger. It inhibits cyclooxygenase-2 (COX-2) and exhibits a range of biological activities including antimicrobial, antioxidant, anti-inflammatory, and anticancer properties. This product is intended for research use only.
  • Natural polyhydroxyphenolic compound
  • Acts as a free radical scavenger
  • Inhibits cyclooxygenase-2 (COX-2)
  • Exhibits antimicrobial activity
  • Provides antioxidant properties
  • Demonstrates anti-inflammatory effects
  • Possesses anticancer activities
  • Improves glucose tolerance in vivo
  • Reduces serum triglyceride concentration in vivo
  • Intended for research use only