Learn More
Medchemexpress LLC Benzoic acid, 3,4,5-trihydroxy-, propyl ester | 121-79-9 | MFCD00002196 | 100.0% | 212.20 | C10H12O5 | 10 MG
SDP
Catalog No. 5000299016
Encompass_Preferred
Propyl gallate (CAS 121-79-9) is an analytical reference standard of the antioxidant propyl gallate, supplied as a solid, white to off-white powder for research and analytical applications. It is used for calibration, method validation, and as a reference material in quantitative assays.
  • Analytical standard for calibration and method validation.
  • High purity suitable for quantitative analysis.
  • Solid, white to off-white form for straightforward handling.
  • Available in small 10 mg quantities for analytical workflows.
  • Includes documentation such as certificate of analysis and safety data sheet for traceability.
Catalog No. 50-002-99016 Supplier Medchemexpress LLC Supplier No. HYN0524R10MG
Propyl gallate (CAS 121-79-9) is an analytical reference standard of the antioxidant propyl gallate, supplied as a solid, white to off-white powder for research and analytical applications. It is used for calibration, method validation, and as a reference material in quantitative assays.
  • Analytical standard for calibration and method validation.
  • High purity suitable for quantitative analysis.
  • Solid, white to off-white form for straightforward handling.
  • Available in small 10 mg quantities for analytical workflows.
  • Includes documentation such as certificate of analysis and safety data sheet for traceability.