Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 3,6-dichloro-2-(methoxy-d3) | 349553-95-3 | 99.7% | 10MG
SDP

Catalog No. 5000231058
Encompass_Preferred

Benzoic acid, 3,6-dichloro-2-(methoxy-d3) | 349553-95-3 | 99.7% | 10MG

Catalog No. 50-002-31058 Supplier Medchemexpress LLC Supplier No. HY121267S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzoic acid, 3,6-dichloro-2-(methoxy-d3) | 349553-95-3 | 99.7% | 10MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.