Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 3,6-dichloro-2-(methoxy-d3)- | 349553-95-3 | 99.7% | 5 MG
SDP

Catalog No. 5000232360
Encompass_Preferred

Dicamba-d3 is a deuterium-labeled isotopologue of dicamba supplied for analytical and research use. It is used as a tracer and internal standard for quantitative methods, enabling accurate measurement in NMR, GC-MS, and LC-MS workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (HPLC 99.7%) for accurate quantitation.
  • Molecular weight 224.06 g/mol; formula C8H3D3Cl2O3.
  • Stable when stored as a powder at -20°C.

Catalog No. 50-002-32360 Supplier Medchemexpress LLC Supplier No. HY121267S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dicamba-d3 is a deuterium-labeled isotopologue of dicamba supplied for analytical and research use. It is used as a tracer and internal standard for quantitative methods, enabling accurate measurement in NMR, GC-MS, and LC-MS workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (HPLC 99.7%) for accurate quantitation.
  • Molecular weight 224.06 g/mol; formula C8H3D3Cl2O3.
  • Stable when stored as a powder at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.