Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 3-borono-, 1-ethyl ester. | 4334-87-6 | MFCD02179444 | 99.7% | 194.00 | C9H11BO4 | 10 G
SDP

Catalog No. 5000303155
Encompass_Preferred

3-Ethoxycarbonylphenylboronic acid is a phenylboronic acid derivative used as an organic reagent and building block in life-science and synthetic chemistry research. It is supplied as a white to off-white solid for research use only and is accompanied by batch-level analytical data.

  • High purity: 99.67% by HPLC.
  • Molecular formula: C9H11BO4.
  • Molecular weight: 194.00 g·mol⁻¹.
  • Appearance: white to off-white solid.
  • Supplied with a certificate of analysis providing batch-specific analytical results.
  • Suitable as a building block for organic synthesis and reagent applications.

Catalog No. 50-003-03155 Supplier Medchemexpress LLC Supplier No. HYW01504210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Ethoxycarbonylphenylboronic acid is a phenylboronic acid derivative used as an organic reagent and building block in life-science and synthetic chemistry research. It is supplied as a white to off-white solid for research use only and is accompanied by batch-level analytical data.

  • High purity: 99.67% by HPLC.
  • Molecular formula: C9H11BO4.
  • Molecular weight: 194.00 g·mol⁻¹.
  • Appearance: white to off-white solid.
  • Supplied with a certificate of analysis providing batch-specific analytical results.
  • Suitable as a building block for organic synthesis and reagent applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.