Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 3-borono-, 1-ethyl ester | 4334-87-6 | MFCD02179444 | 99.7% | 194.00 g/mol | C9H11BO4 | 25 G
SDP

Catalog No. 5000303442
Encompass_Preferred

3-Ethoxycarbonylphenylboronic acid is an organoboron reagent used as a building block in organic synthesis and life-science research. It is commonly employed in Suzuki-Miyaura cross-coupling reactions and other boron-mediated transformations. Supplied as a white to off-white solid with verified high purity, it is suitable for research and development applications.

  • Molecular formula: C9H11BO4.
  • Molecular weight: 194.00 g/mol.
  • High purity by HPLC (reported 99.67%).
  • White to off-white solid appearance.
  • Suitable for Suzuki-Miyaura cross-coupling and building-block chemistry.

Catalog No. 50-003-03442 Supplier Medchemexpress LLC Supplier No. 25GHYW015042
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Ethoxycarbonylphenylboronic acid is an organoboron reagent used as a building block in organic synthesis and life-science research. It is commonly employed in Suzuki-Miyaura cross-coupling reactions and other boron-mediated transformations. Supplied as a white to off-white solid with verified high purity, it is suitable for research and development applications.

  • Molecular formula: C9H11BO4.
  • Molecular weight: 194.00 g/mol.
  • High purity by HPLC (reported 99.67%).
  • White to off-white solid appearance.
  • Suitable for Suzuki-Miyaura cross-coupling and building-block chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.