Learn More
Medchemexpress LLC Benzoic acid, 3-phenoxy- | 3739-38-6 | MFCD00002498 | 98.2% | 214.22 g/mol | C13H10O3 | 25 MG
SDP
Catalog No. 5000303044
Encompass_Preferred
3-Phenoxybenzoic acid is an analytical reference standard used in research and analytical testing. It serves as a metabolite marker of pyrethroid insecticides and is supplied as a solid reference material suitable for method development, calibration, and quantitative analysis in environmental, toxicology, and analytical chemistry applications.
  • Analytical reference standard for pyrethroid metabolite studies.
  • High purity suitable for HPLC and quantitative analysis.
  • Molecular formula C13H10O3; molecular weight 214.22 g/mol.
  • Available in small package sizes to support analytical workflows.
  • Batch-specific certificate of analysis available for assay verification.
Catalog No. 50-003-03044 Supplier Medchemexpress LLC Supplier No. 25MGHYW014225R
Add to Cart
Add to Cart
3-Phenoxybenzoic acid is an analytical reference standard used in research and analytical testing. It serves as a metabolite marker of pyrethroid insecticides and is supplied as a solid reference material suitable for method development, calibration, and quantitative analysis in environmental, toxicology, and analytical chemistry applications.
  • Analytical reference standard for pyrethroid metabolite studies.
  • High purity suitable for HPLC and quantitative analysis.
  • Molecular formula C13H10O3; molecular weight 214.22 g/mol.
  • Available in small package sizes to support analytical workflows.
  • Batch-specific certificate of analysis available for assay verification.