Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 3-phenoxy- | 3739-38-6 | MFCD00002498 | 99.6% | 214.22 g/mol | C13H10O3 | 50 G
SDP

Catalog No. 5000304125
Encompass_Preferred

3-Phenoxybenzoic acid (3-PBA) is a metabolite of pyrethroid insecticides commonly used as an analytical standard and research reagent. It is employed in studies of immunotoxicity, oxidative stress, and environmental/biological monitoring, and is supplied with high purity suitable for analytical applications.

  • Metabolite of pyrethroid insecticides used as an analytical standard.
  • Chemical formula C13H10O3; molecular weight 214.22 g/mol.
  • High purity suitable for analytical work (≈99.6%).
  • Available in multiple solid pack sizes up to 50 g and as a solution.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for research and analytical applications; not for human therapeutic use.

Catalog No. 50-003-04125 Supplier Medchemexpress LLC Supplier No. HYW01422550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-Phenoxybenzoic acid (3-PBA) is a metabolite of pyrethroid insecticides commonly used as an analytical standard and research reagent. It is employed in studies of immunotoxicity, oxidative stress, and environmental/biological monitoring, and is supplied with high purity suitable for analytical applications.

  • Metabolite of pyrethroid insecticides used as an analytical standard.
  • Chemical formula C13H10O3; molecular weight 214.22 g/mol.
  • High purity suitable for analytical work (≈99.6%).
  • Available in multiple solid pack sizes up to 50 g and as a solution.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for research and analytical applications; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.