Learn More
Medchemexpress LLC Benzoic acid, 4-[[(2-amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]- | 119-24-4 | 98.1% | C14H12N6O3 | 10 G
SDP
Catalog No. 5000434425
Encompass_Preferred
Pteroic acid is a precursor to significant compounds like Folic acid and Vitamin B12. It facilitates the examination of how different compounds influence cell growth and development. This product is for research use only and not sold to patients.
  • Precursor to Folic acid and Vitamin B12
  • Facilitates examination of how compounds influence cell growth and development
  • Exhibits inhibitory activity against CDK4 complex in Sf9 cells
  • Enhances activity of partially purified beef liver folylpolyglutamate synthetase (FPGS)
Catalog No. 50-004-34425 Supplier Medchemexpress LLC Supplier No. HY7913910G
Add to Cart
Add to Cart
Pteroic acid is a precursor to significant compounds like Folic acid and Vitamin B12. It facilitates the examination of how different compounds influence cell growth and development. This product is for research use only and not sold to patients.
  • Precursor to Folic acid and Vitamin B12
  • Facilitates examination of how compounds influence cell growth and development
  • Exhibits inhibitory activity against CDK4 complex in Sf9 cells
  • Enhances activity of partially purified beef liver folylpolyglutamate synthetase (FPGS)