Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 4-[(3aS,4R,9bR)-6-ethoxy-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-4-yl]- | 1357248-83-9 | 98.7% | 335.40 | 5 MG
SDP

Catalog No. 5000364228
Encompass_Preferred

Benzoic acid, 4-[(3aS,4R,9bR)-6-ethoxy-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-4-yl]- | 1357248-83-9 | 98.7% | 335.40 | 5 MG

Catalog No. 50-003-64228 Supplier Medchemexpress LLC Supplier No. 5MGHY123905
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzoic acid, 4-[(3aS,4R,9bR)-6-ethoxy-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-4-yl]- | 1357248-83-9 | 98.7% | 335.40 | 5 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.