Learn More
Medchemexpress LLC Benzoic acid, 4-[4-[[(3R)-1-butyl-3-[(R)-cyclohexylhydroxymethyl]-2,5-dioxo-1,4,9-triazaspiro [5.5]undec-9-yl]methyl]phenoxy]-, hydrochloride (1:1) | 461023-63-2 | 99.9% | 614.17 | 25 MG
SDP
Catalog No. 5000384286
Encompass_Preferred
Aplaviroc hydrochloride is a CCR5 antagonist derived from SDP, with IC50s ranging from 0.1-0.4 nM for HIV-1Ba-L, HIV-1JRFL, and HIV-1MOKW. It is intended for research use only.
  • Target: CCR5 antagonist
  • Potency: Potent activity against wild-type R5 HIV-1 strains with IC50 values of 0.1 to 0.4 nM
  • Mechanism: Suppresses infectivity and replication of HIV-1MDR variants at low concentrations (IC50 values of 0.4 to 0.6 nM)
  • Binding Affinity: Binds to CCR5 with high affinity (Kd value of 2.9±1.0 nM)
  • Inhibition: Potently blocks rgp120/sCD4 binding to CCR5 with an IC50 value of 2.7 nM
  • In Vivo Efficacy: Suppresses R5 HIV-1 viremia in hu-PBMC-NOG mice at a dosage of 60 mg/kg
Catalog No. 50-003-84286 Supplier Medchemexpress LLC Supplier No. 25MGHY17450A
Aplaviroc hydrochloride is a CCR5 antagonist derived from SDP, with IC50s ranging from 0.1-0.4 nM for HIV-1Ba-L, HIV-1JRFL, and HIV-1MOKW. It is intended for research use only.
  • Target: CCR5 antagonist
  • Potency: Potent activity against wild-type R5 HIV-1 strains with IC50 values of 0.1 to 0.4 nM
  • Mechanism: Suppresses infectivity and replication of HIV-1MDR variants at low concentrations (IC50 values of 0.4 to 0.6 nM)
  • Binding Affinity: Binds to CCR5 with high affinity (Kd value of 2.9±1.0 nM)
  • Inhibition: Potently blocks rgp120/sCD4 binding to CCR5 with an IC50 value of 2.7 nM
  • In Vivo Efficacy: Suppresses R5 HIV-1 viremia in hu-PBMC-NOG mice at a dosage of 60 mg/kg