Learn More
Medchemexpress LLC Benzoic acid, 4-[5-(4,7-dimethyl-2-benzofuranyl)-1,2,4-oxadiazol-3-yl]- | 1952276-71-9 | 98.9% | 334.33 | 50 MG
SDP
Catalog No. 5000439664
Encompass_Preferred
KCL-286 (C286) is an orally active and brain-penetrant retinoic acid receptor (RAR) β2 agonist (EC50 = 1.9 nM). It activates RARβ2 in injured neurons, inducing axonal regeneration and modulating neuroinflammation and extracellular matrix molecules. It can also modulate the expression of chondroitin sulfate proteoglycans (CSPGs) by neuronal secretion of decorin, promoting myelination and aiding axonal growth.
  • Targets RARβ2 with selectivity over RAR α and RAR γ.
  • Activates RARβ2 in injured neurons.
  • Induces axonal regeneration of spinal and sensory nerves.
  • Modulates neuroinflammation and extracellular matrix molecules.
  • Promotes myelination and aids axonal growth.
  • Suitable for research in spinal cord injury and traumatic nerve injury.
Catalog No. 50-004-39664 Supplier Medchemexpress LLC Supplier No. HY11157350MG
KCL-286 (C286) is an orally active and brain-penetrant retinoic acid receptor (RAR) β2 agonist (EC50 = 1.9 nM). It activates RARβ2 in injured neurons, inducing axonal regeneration and modulating neuroinflammation and extracellular matrix molecules. It can also modulate the expression of chondroitin sulfate proteoglycans (CSPGs) by neuronal secretion of decorin, promoting myelination and aiding axonal growth.
  • Targets RARβ2 with selectivity over RAR α and RAR γ.
  • Activates RARβ2 in injured neurons.
  • Induces axonal regeneration of spinal and sensory nerves.
  • Modulates neuroinflammation and extracellular matrix molecules.
  • Promotes myelination and aids axonal growth.
  • Suitable for research in spinal cord injury and traumatic nerve injury.