Learn More
Medchemexpress LLC Benzoic acid, 4-(6-fluoro-3,5-dihydro-3-oxo-2H-pyrazolo[4,3-c]quinolin-2-yl) | 486449-65-4 | 98.0% | 323.28 g/mol | C17H10FN3O3 | 5 MG
SDP
Catalog No. 5000213024
Encompass_Preferred
CD80-IN-3 is a small-molecule inhibitor of the immune co-stimulatory protein CD80 that disrupts CD80-CD28 interaction. It is reported to have an EC50 of 630 nM and a binding Kd of 125 nM, and is supplied as a high-purity solid for in vitro research applications.
  • Potent inhibition of CD80-CD28 interaction (EC50 630 nM; Kd 125 nM).
  • Well-characterized small molecule: MW 323.28, formula C17H10FN3O3.
  • High purity suitable for biochemical and cell-based assays.
  • Available in small research quantities for screening and profiling.
  • Stable powder with defined storage conditions for long-term retention.
  • Light brown to yellow solid, convenient for formulation in DMSO.
Catalog No. 50-002-13024 Supplier Medchemexpress LLC Supplier No. HY1008915MG
CD80-IN-3 is a small-molecule inhibitor of the immune co-stimulatory protein CD80 that disrupts CD80-CD28 interaction. It is reported to have an EC50 of 630 nM and a binding Kd of 125 nM, and is supplied as a high-purity solid for in vitro research applications.
  • Potent inhibition of CD80-CD28 interaction (EC50 630 nM; Kd 125 nM).
  • Well-characterized small molecule: MW 323.28, formula C17H10FN3O3.
  • High purity suitable for biochemical and cell-based assays.
  • Available in small research quantities for screening and profiling.
  • Stable powder with defined storage conditions for long-term retention.
  • Light brown to yellow solid, convenient for formulation in DMSO.