Learn More
Medchemexpress LLC Benzoic acid, 4-amino- | 150-13-0 | 98.8% | 1 ML
SDP
Catalog No. 5000407543
Encompass_Preferred
4-Aminobenzoic acid (PABA) is a chemical compound that serves as an intermediate in the synthesis of folic acid by bacteria, plants, and fungi. It exhibits antibacterial and plant growth regulating properties, making it valuable for various research applications. This product is intended for research use only.
  • Intermediate in folic acid synthesis by bacteria, plants, and fungi.
  • Possesses antibacterial properties.
  • Regulates plant growth.
  • Acts as a scavenger of reactive oxygen species, useful in sunscreen applications.
  • Promotes plant resistance against bacterial and viral pathogens.
  • Supplied as a 10 mM solution in DMSO.
Catalog No. 50-004-07543 Supplier Medchemexpress LLC Supplier No. HYB100810MM/1ML
4-Aminobenzoic acid (PABA) is a chemical compound that serves as an intermediate in the synthesis of folic acid by bacteria, plants, and fungi. It exhibits antibacterial and plant growth regulating properties, making it valuable for various research applications. This product is intended for research use only.
  • Intermediate in folic acid synthesis by bacteria, plants, and fungi.
  • Possesses antibacterial properties.
  • Regulates plant growth.
  • Acts as a scavenger of reactive oxygen species, useful in sunscreen applications.
  • Promotes plant resistance against bacterial and viral pathogens.
  • Supplied as a 10 mM solution in DMSO.