Learn More
Medchemexpress LLC Benzoic acid, 4-[(aminoiminomethyl)amino]-, 4-[[2-(2,5-dioxo-1-pyrrolidinyl)ethyl]thio] phenyl ester | 114568-26-2 | 98.1% | 412.46 | 100 MG
SDP
Catalog No. 5000436960
Encompass_Preferred
Patamostat (E-3123) is a potent protease inhibitor that effectively inhibits trypsin, plasmin, and thrombin with IC50 values of 39 nM, 950 nM, and 1.9 μM, respectively. It may have suppressing effects on the pathogenesis and development of acute pancreatitis. This product is for research use only and is not sold to patients.
  • Potent protease inhibitor.
  • Inhibits trypsin.
  • Inhibits plasmin.
  • Inhibits thrombin.
  • May suppress the pathogenesis and development of acute pancreatitis.
Catalog No. 50-004-36960 Supplier Medchemexpress LLC Supplier No. HY114080100MG
Patamostat (E-3123) is a potent protease inhibitor that effectively inhibits trypsin, plasmin, and thrombin with IC50 values of 39 nM, 950 nM, and 1.9 μM, respectively. It may have suppressing effects on the pathogenesis and development of acute pancreatitis. This product is for research use only and is not sold to patients.
  • Potent protease inhibitor.
  • Inhibits trypsin.
  • Inhibits plasmin.
  • Inhibits thrombin.
  • May suppress the pathogenesis and development of acute pancreatitis.