Learn More
Medchemexpress LLC Benzoic acid, 4-bromo-2-methyl- | 68837-59-2 | 215.046 | 25 G
SDP
Catalog No. 5000449858
Encompass_Preferred
4-Bromo-2-methylbenzoic acid is an important compound that can be used in the synthesis of other active compounds. It can be utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate. This product is for research use only and has not been fully validated for medical applications.
  • Can be used in the synthesis of other active compounds.
  • Utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate.
  • For research use only.
  • Appears as an off-white to pink solid.
  • Purity verified by HPLC at 99.51%.
Catalog No. 50-004-49858 Supplier Medchemexpress LLC Supplier No. 25GHYW004863
4-Bromo-2-methylbenzoic acid is an important compound that can be used in the synthesis of other active compounds. It can be utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate. This product is for research use only and has not been fully validated for medical applications.
  • Can be used in the synthesis of other active compounds.
  • Utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate.
  • For research use only.
  • Appears as an off-white to pink solid.
  • Purity verified by HPLC at 99.51%.