Learn More
Medchemexpress LLC Benzoic acid, 4-[[(cyclopropylmethyl)[4-(2-fluorophenoxy)benzoyl]amino]methyl] | 1664335-55-0 | 99.0% | 419.44 g/mol | C25H22FNO4 | 1 ML
SDP
Catalog No. 5000307969
Encompass_Preferred
TAK-615 is a small-molecule negative allosteric modulator of the lysophosphatidic acid receptor 1 (LPA1), supplied as a 10 mM solution in 1 mL for research use. It is commonly used in studies of pulmonary fibrosis and LPA1-mediated signaling. CAS 1664335-55-0; molecular weight 419.44 g/mol.
  • Negative allosteric modulator of the LPA1 receptor for receptor pharmacology studies.
  • Supplied as a 10 mM solution in 1 mL for ready-to-use in vitro experiments.
  • High purity (98.97%) suitable for research applications.
  • Chemical formula C25H22FNO4 and molecular weight 419.44 g/mol.
  • Provided with certificate of analysis for batch-specific quality data.
Catalog No. 50-003-07969 Supplier Medchemexpress LLC Supplier No. HY11795910MM 1ML
TAK-615 is a small-molecule negative allosteric modulator of the lysophosphatidic acid receptor 1 (LPA1), supplied as a 10 mM solution in 1 mL for research use. It is commonly used in studies of pulmonary fibrosis and LPA1-mediated signaling. CAS 1664335-55-0; molecular weight 419.44 g/mol.
  • Negative allosteric modulator of the LPA1 receptor for receptor pharmacology studies.
  • Supplied as a 10 mM solution in 1 mL for ready-to-use in vitro experiments.
  • High purity (98.97%) suitable for research applications.
  • Chemical formula C25H22FNO4 and molecular weight 419.44 g/mol.
  • Provided with certificate of analysis for batch-specific quality data.