Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 4-formyl-, methyl ester | 1571-08-0 | 100.0% | C9H8O3 | 250 G
SDP

Catalog No. 5000358661
Encompass_Preferred

Methyl 4-formylbenzoate is an intermediate that can be used for the synthesis of chalcone amide α-glucosidase inhibitors. It appears as a white to off-white solid.

  • Used for the synthesis of chalcone amide α-glucosidase inhibitors.
  • Appears as a white to off-white solid.
  • Molecular weight is 164.16.
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years.
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-58661 Supplier Medchemexpress LLC Supplier No. 250GHY78000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl 4-formylbenzoate is an intermediate that can be used for the synthesis of chalcone amide α-glucosidase inhibitors. It appears as a white to off-white solid.

  • Used for the synthesis of chalcone amide α-glucosidase inhibitors.
  • Appears as a white to off-white solid.
  • Molecular weight is 164.16.
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years.
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.