Learn More
Medchemexpress LLC Benzoic acid, 4-formyl-, methyl ester | 1571-08-0 | 99.99% | C9H8O3 | 25 G
SDP
Catalog No. 5000359148
Encompass_Preferred
Methyl 4-formylbenzoate is an intermediate used for the synthesis of chalcone amide α-glucosidase inhibitors. It has a molecular weight of 164.16 and a chemical formula of C9H8O3, appearing as a white to off-white solid.
  • Purity of 99.99%.
  • Intended for research purposes only.
  • Useful as an intermediate for synthesizing chalcone amide α-glucosidase inhibitors.
Catalog No. 50-003-59148 Supplier Medchemexpress LLC Supplier No. 25GHY78000
Methyl 4-formylbenzoate is an intermediate used for the synthesis of chalcone amide α-glucosidase inhibitors. It has a molecular weight of 164.16 and a chemical formula of C9H8O3, appearing as a white to off-white solid.
  • Purity of 99.99%.
  • Intended for research purposes only.
  • Useful as an intermediate for synthesizing chalcone amide α-glucosidase inhibitors.