Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 4-hydroxy-3,5-dimethoxy- | 530-57-4 | 232023 | 99.9% | 198.17 | 1 ML
SDP

Catalog No. 5000427209
Encompass_Preferred

Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation. This product is intended for research and analytical applications. It is supplied as a white to off-white solid.

  • Target is endogenous metabolite and bacterial
  • Research areas include metabolic disease
  • Purity (HPLC): 99.85%
  • Storage as powder: -20°C for 3 years; 4°C for 2 years
  • Storage in solvent: -80°C for 2 years; -20°C for 1 year

Catalog No. 50-004-27209 Supplier Medchemexpress LLC Supplier No. HYN033910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Syringic acid is correlated with high antioxidant activity and inhibition of LDL oxidation. This product is intended for research and analytical applications. It is supplied as a white to off-white solid.

  • Target is endogenous metabolite and bacterial
  • Research areas include metabolic disease
  • Purity (HPLC): 99.85%
  • Storage as powder: -20°C for 3 years; 4°C for 2 years
  • Storage in solvent: -80°C for 2 years; -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.