Learn More
Medchemexpress LLC Benzoic acid, 4-hydroxy-3-methoxy-, ethyl ester | 617-05-0 | 99.87% | 25 G
SDP
Catalog No. 5000414502
Encompass_Preferred
Ethyl Vanillate is a fungicidal agent that inhibits 17β-HSD2 with an IC50 of 1.3 μM. It is for research use only.
  • Fungicidal agent
  • Inhibits 17β-HSD2
Catalog No. 50-004-14502 Supplier Medchemexpress LLC Supplier No. 25GHYB1643
Ethyl Vanillate is a fungicidal agent that inhibits 17β-HSD2 with an IC50 of 1.3 μM. It is for research use only.
  • Fungicidal agent
  • Inhibits 17β-HSD2