Learn More
Medchemexpress LLC Benzoic acid, 4-(hydroxymethyl)- | 3006-96-0 | MFCD00017598 | 99.9% | 152.15 g/mol | C8H8O3 | 100 G
SDP
Catalog No. 5000299777
Encompass_Preferred
4-(Hydroxymethyl)benzoic acid (CAS 3006-96-0) is a benzoic acid derivative used as a biochemical reagent and building block in organic synthesis, assay development, and life-science research. Molecular formula C8H8O3; molecular weight 152.15 g/mol. Purity and appearance vary by batch - consult the certificate of analysis for batch-specific data.
  • Used as a building block for synthesis and conjugation.
  • Suitable for biochemical reagent and assay development.
  • Provided as a solid for convenient storage and handling.
  • Documented molecular formula C8H8O3 and molecular weight 152.15 g/mol.
  • High purity available; consult batch-specific COA for exact value.
  • Available in multiple package sizes including 100 g.
Catalog No. 50-002-99777 Supplier Medchemexpress LLC Supplier No. HYW017384100G
4-(Hydroxymethyl)benzoic acid (CAS 3006-96-0) is a benzoic acid derivative used as a biochemical reagent and building block in organic synthesis, assay development, and life-science research. Molecular formula C8H8O3; molecular weight 152.15 g/mol. Purity and appearance vary by batch - consult the certificate of analysis for batch-specific data.
  • Used as a building block for synthesis and conjugation.
  • Suitable for biochemical reagent and assay development.
  • Provided as a solid for convenient storage and handling.
  • Documented molecular formula C8H8O3 and molecular weight 152.15 g/mol.
  • High purity available; consult batch-specific COA for exact value.
  • Available in multiple package sizes including 100 g.