Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzoic acid, 4-methyl-3-nitro- | 96-98-0 | MFCD00007174 | 100.0% | 181.15 g/mol | C8H7NO4 | 25 G
SDP

Catalog No. 5000302848
Encompass_Preferred

4-Methyl-3-nitrobenzoic acid (CAS 96-98-0) is an organic biochemical reagent used as a building block and intermediate in medicinal chemistry and organic synthesis. Supplied as an off-white to light yellow solid with a high reported purity, it is intended for laboratory research and analytical applications. Follow supplier-recommended storage conditions for powder and solutions to maintain stability.

  • High reported purity (99.96%).
  • Molecular weight 181.15 g/mol.
  • Chemical formula C8H7NO4.
  • Solid, off-white to light yellow appearance.
  • Suitable as a synthetic intermediate for research chemistry.
  • Storage recommendations provided for powder and in-solution stability.

Catalog No. 50-003-02848 Supplier Medchemexpress LLC Supplier No. 25GHYW021293
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Methyl-3-nitrobenzoic acid (CAS 96-98-0) is an organic biochemical reagent used as a building block and intermediate in medicinal chemistry and organic synthesis. Supplied as an off-white to light yellow solid with a high reported purity, it is intended for laboratory research and analytical applications. Follow supplier-recommended storage conditions for powder and solutions to maintain stability.

  • High reported purity (99.96%).
  • Molecular weight 181.15 g/mol.
  • Chemical formula C8H7NO4.
  • Solid, off-white to light yellow appearance.
  • Suitable as a synthetic intermediate for research chemistry.
  • Storage recommendations provided for powder and in-solution stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.