Learn More
Medchemexpress LLC Benzoic acid, 5-[2-(2,5-dimethoxyphenyl)ethyl]-2-hydroxy-, methyl ester | 150779-71-8 | MFCD00920170 | 98.0% | 316.35 g/mol | C18H20O5 | 25 MG
SDP
Catalog No. 5000245380
Encompass_Preferred
SDZ281-977 is a small-molecule derivative of the epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor lavendustin A. Supplied as a high-purity solid for research use, it is used in biochemical and cell-based studies to probe EGFR signaling and kinase activity.
  • High purity (98.0%).
  • Chemical formula C18H20O5; molecular weight 316.35 g/mol.
  • Solid powder form suitable for storage and analytical use.
  • Stable under recommended storage: powder at -20°C (long term) and 4°C (short term); in solvent stability, -80°C for short-term storage.
  • Supplied in 25 mg quantity for small-scale research applications.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-45380 Supplier Medchemexpress LLC Supplier No. HY10175625MG
SDZ281-977 is a small-molecule derivative of the epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor lavendustin A. Supplied as a high-purity solid for research use, it is used in biochemical and cell-based studies to probe EGFR signaling and kinase activity.
  • High purity (98.0%).
  • Chemical formula C18H20O5; molecular weight 316.35 g/mol.
  • Solid powder form suitable for storage and analytical use.
  • Stable under recommended storage: powder at -20°C (long term) and 4°C (short term); in solvent stability, -80°C for short-term storage.
  • Supplied in 25 mg quantity for small-scale research applications.
  • Intended for research use only; not for clinical or human use.