Learn More
Medchemexpress LLC Benzoic acid, 5-[(3-carboxy-5-methyl-4-oxo-2,5-cyclohexadien-1-ylidene)(2,6-dichloro-3-sulfophenyl)methyl]-2-hydroxy-3-methyl-, sodium salt (1:3) | 1667-99-8 | 50 G
SDP
Catalog No. 5000410900
Encompass_Preferred
Chromeazurol S, an azo dye, is frequently utilized as an indicator in analytical chemistry for detecting metal ions such as copper, nickel, and cobalt. It facilitates their detection and quantification by changing from yellow to blue in their presence. This compound can be applied to a test strip or added directly to a solution for analysis. As a biochemical reagent, it is suitable for use as a biological material or organic compound in life science-related research.
  • Used as an indicator in analytical chemistry
  • Detects metal ions like copper, nickel, and cobalt
  • Changes from yellow to blue in the presence of metal ions
  • Can be applied to test strips or added directly to solutions
  • Suitable for life science-related research
Catalog No. 50-004-10900 Supplier Medchemexpress LLC Supplier No. HYD080050G
Add to Cart
Add to Cart
Chromeazurol S, an azo dye, is frequently utilized as an indicator in analytical chemistry for detecting metal ions such as copper, nickel, and cobalt. It facilitates their detection and quantification by changing from yellow to blue in their presence. This compound can be applied to a test strip or added directly to a solution for analysis. As a biochemical reagent, it is suitable for use as a biological material or organic compound in life science-related research.
  • Used as an indicator in analytical chemistry
  • Detects metal ions like copper, nickel, and cobalt
  • Changes from yellow to blue in the presence of metal ions
  • Can be applied to test strips or added directly to solutions
  • Suitable for life science-related research