Learn More
U.S. Pharmacopeia Benzoic Acid, 65-85-0, MFCD00002398, 300mg
Catalog No. NC9569831
Encompass
Molecular Formula C7H6O2, Molecular Weight 122.12, Melting Point 249.8 - 255.2 °F (121 - 124 °C), Boiling Point 480.2 °F (249 °C), Synonyms: Benzene carboxylic acid
Catalog No. NC9569831 Supplier U.S. Pharmacopeia Supplier No. 1055002
Molecular Formula C7H6O2, Molecular Weight 122.12, Melting Point 249.8 - 255.2 °F (121 - 124 °C), Boiling Point 480.2 °F (249 °C), Synonyms: Benzene carboxylic acid