Learn More
Cambridge Isotope Laboratories BENZOIC ACID CALIBRANT FOR QNMR 5mM BENZOIC ACID IN DMSO-D6 (120 UL FILL VOLUME), 1 3MM TUBE
Catalog No. 501631347
Encompass
BENZOIC ACID CALIBRANT FOR QNMR 5mM BENZOIC ACID IN DMSO-D6 (120 UL FILL VOLUME), 1 3MM TUBE
Catalog No. 50-163-1347 Supplier Cambridge Isotope Laboratories Supplier No. DLM9491E
BENZOIC ACID CALIBRANT FOR QNMR 5mM BENZOIC ACID IN DMSO-D6 (120 UL FILL VOLUME), 1 3MM TUBE