Learn More
Apexbio Technology LLC Benzoic acid lithium 553-54-8 500g
SDP
Catalog No. 5000094512
Encompass_Preferred
Lithium benzoate (553-54-8) is a small-molecule inhibitor It exerts its biological activity primarily through disruption of microbial cell membrane integrity and inhibition of microbial enzyme function In in vitro studies lithium benzoate demonstrates antimicrobial activity with reported inhibitory concentrations (IC50) ranging from micromolar to millimolar against common microbial strains depending on organism sensitivity and experimental conditions Based on these pharmacological properties lithium benzoate holds research potential in pharmacological formulation development cell culture medium preservation and microbial contamination control studies in biomedical laboratories
Catalog No. 50-000-94512 Supplier Apexbio Technology LLC Supplier No. A9991500000
Lithium benzoate (553-54-8) is a small-molecule inhibitor It exerts its biological activity primarily through disruption of microbial cell membrane integrity and inhibition of microbial enzyme function In in vitro studies lithium benzoate demonstrates antimicrobial activity with reported inhibitory concentrations (IC50) ranging from micromolar to millimolar against common microbial strains depending on organism sensitivity and experimental conditions Based on these pharmacological properties lithium benzoate holds research potential in pharmacological formulation development cell culture medium preservation and microbial contamination control studies in biomedical laboratories