Learn More
Sigma Aldrich Fine Chemicals Biosciences Benzoic acid NISTR TM34
Catalog No. 502908341
Encompass
This Standard Reference Material (SRM) consists of purified benzoic acid (C6H5COOH). SRM 350c is intended for use in acidimetric standardization. This material is NOT suitable for use as a standard in quantitative nuclear magneticresonance spectrometry (qNMR). A unit of SRM 350c consists of 30 g of benzoic acid.
Catalog No. 50-290-8341 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST350C
This Standard Reference Material (SRM) consists of purified benzoic acid (C6H5COOH). SRM 350c is intended for use in acidimetric standardization. This material is NOT suitable for use as a standard in quantitative nuclear magneticresonance spectrometry (qNMR). A unit of SRM 350c consists of 30 g of benzoic acid.