Learn More
GFS Chemicals Inc BENZOIC ACID, PRIMARY STANDARD | 25 G | GFS CHEMICALS, INC. Item #: 15312 | Bulk #: 1681
SDP
Catalog No. NC1920305
Encompass_Preferred
BENZOIC ACID, PRIMARY STANDARD | Chemical Formula : C!6H!5COOH | CAS # : 65-85-0 | Specifications : Assay 99.95 - 100.05 %, Chlorine compounds (%) 0.005 % , Freezing point (deg C) 122 - 123 C, Heavy metals (Pb) 0.0005 % , Insoluble in methanol (%) 0.005 % , NIST Traceable, Residue after ignition 0.005 % , Substances reducing permanganate, Sulfur compounds 0.002 %
Catalog No. NC1920305 Supplier GFS Chemicals Inc Supplier No. 15312
BENZOIC ACID, PRIMARY STANDARD | Chemical Formula : C!6H!5COOH | CAS # : 65-85-0 | Specifications : Assay 99.95 - 100.05 %, Chlorine compounds (%) 0.005 % , Freezing point (deg C) 122 - 123 C, Heavy metals (Pb) 0.0005 % , Insoluble in methanol (%) 0.005 % , NIST Traceable, Residue after ignition 0.005 % , Substances reducing permanganate, Sulfur compounds 0.002 %