Learn More
Medchemexpress LLC Benzoic acid (Standard) | 65-85-0 | 99.7% | 122.12 | 100 MG
SDP
Catalog No. 5000427986
Encompass_Preferred
Benzoic acid is an analytical standard intended for research and analytical applications. It is an aromatic alcohol naturally found in various plants and widely used as an additive in food, drinks, cosmetics, and other products, functioning as a preservative by inhibiting both bacteria and fungi.
  • Analytical standard for research and analysis
  • Naturally occurring aromatic alcohol
  • Preservative action by inhibiting bacteria and fungi
  • Commonly used in HPLC, GC, and MS experiments
Catalog No. 50-004-27986 Supplier Medchemexpress LLC Supplier No. HYN0216R100MG
Benzoic acid is an analytical standard intended for research and analytical applications. It is an aromatic alcohol naturally found in various plants and widely used as an additive in food, drinks, cosmetics, and other products, functioning as a preservative by inhibiting both bacteria and fungi.
  • Analytical standard for research and analysis
  • Naturally occurring aromatic alcohol
  • Preservative action by inhibiting bacteria and fungi
  • Commonly used in HPLC, GC, and MS experiments