Learn More
Medchemexpress LLC Benzoic anhydride | 93-97-0 | MFCD00003073 | 1mL
SDP
Catalog No. 5000267980
Encompass_Preferred
Benzoic anhydride is a catalyst that can be used for organic synthesis reactions Benzoic anhydride is used as a chemical titration reagent for selective quantification and deactivation of phenolic hydroxyl groups on the surface of carbon nanotubes[1][2]
Catalog No. 50-002-67980 Supplier Medchemexpress LLC Supplier No. HYY137410MM/1ML
Benzoic anhydride is a catalyst that can be used for organic synthesis reactions Benzoic anhydride is used as a chemical titration reagent for selective quantification and deactivation of phenolic hydroxyl groups on the surface of carbon nanotubes[1][2]