Learn More
Medchemexpress LLC Benzoic-carboxy-13C acid, 2-methyl- | 70838-82-3 | 99% | 137.14 | 25 MG
SDP
Catalog No. 5000402325
Encompass_Preferred
o-Toluic acid-13C is the 13C labeled version of o-Toluic acid (2-Methylbenzoic acid), which is a benzoic acid substituted with a methyl group at position 2. It functions as a xenobiotic metabolite. This product is intended for research use only and is not sold to patients.
  • 13C labeled o-Toluic acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like 13C can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-02325 Supplier Medchemexpress LLC Supplier No. 25MGHY41494S1
o-Toluic acid-13C is the 13C labeled version of o-Toluic acid (2-Methylbenzoic acid), which is a benzoic acid substituted with a methyl group at position 2. It functions as a xenobiotic metabolite. This product is intended for research use only and is not sold to patients.
  • 13C labeled o-Toluic acid.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like 13C can affect the pharmacokinetic and metabolic profiles of drugs.