Learn More
Medchemexpress LLC Benzoin | 119-53-9 | 99.95% | 1000 G
SDP
Catalog No. 5000411385
Encompass_Preferred
Benzoin (DL-Benzoin) is a PI3Kα inhibitor with anticancer effects. It inhibits the growth of the colon cancer cell line (HCT-116) and exhibits antiproliferative activities against HCT-116 cells with an IC50 of 2.12 mM.
  • Pi3Kα inhibitor
  • Anticancer effects
  • Antiproliferative activities
  • Inhibits growth of HCT-116 colon cancer cell line
Catalog No. 50-004-11385 Supplier Medchemexpress LLC Supplier No. HYB15501000G
Benzoin (DL-Benzoin) is a PI3Kα inhibitor with anticancer effects. It inhibits the growth of the colon cancer cell line (HCT-116) and exhibits antiproliferative activities against HCT-116 cells with an IC50 of 2.12 mM.
  • Pi3Kα inhibitor
  • Anticancer effects
  • Antiproliferative activities
  • Inhibits growth of HCT-116 colon cancer cell line