Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Benzoin Ethyl Ether (ethoxy phenylacetophenone) 50 GR
SDP

Catalog No. 50980198
Encompass_Preferred

(ethoxy phenylacetophenone)
CAS #574-09-4 C₆H₅COCH(CO₂H₅)C₆H₅ F.W. 240.31
Melting point: 56-59°C (133-138 °F)

Catalog No. 50-980-198 Supplier Electron Microscopy Sciences Supplier No. 11280
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(ethoxy phenylacetophenone)
CAS #574-09-4 C₆H₅COCH(CO₂H₅)C₆H₅ F.W. 240.31
Melting point: 56-59°C (133-138 °F)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.