Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoin Methyl Ether 96%, Thermo Scientific™

Catalog No. AC105521000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105521000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105521000 Supplier Thermo Scientific Chemicals Supplier No. 105521000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3524-62-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
InChI Key BQZJOQXSCSZQPS-UHFFFAOYSA-N
Synonym benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
PubChem CID 98097
IUPAC Name 2-methoxy-1,2-diphenylethanone
SMILES COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 45°C to 49°C
Color Green to White
Infrared Spectrum Authentic
Linear Formula C6H5CH(OCH3)COC6H5
Quantity 100 g
Beilstein 08, 174
Merck Index 10, 1095
Formula Weight 226.27
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Benzoin methyl ether, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.