Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Benzoin, U.V. Catalyst 100 GR
SDP

Catalog No. 50980197
Encompass_Preferred

CAS #119-53-9 C₆H₅CH(OH)COC₆H₅ F.W. 212.25
Melting point: 134-136°C

Catalog No. 50-980-197 Supplier Electron Microscopy Sciences Supplier No. 11270
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #119-53-9 C₆H₅CH(OH)COC₆H₅ F.W. 212.25
Melting point: 134-136°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.