Learn More
AdipoGen Benzokfluoranthene
SDP
Supplier:  AdipoGen CDXB0452M100
Encompass_Preferred
Chemical. CAS 207-08-9. Formula C20H12. Molecular Weight 252.31. Benzokfluoranthene is an optical sensor for nitro-aromatic compounds, chosen due to fluorescence quenching of the molecule see Reference 2. Polycyclic aromatic hydrocarbon PAH compound. Benzokfluoranthene is primarily found in gasoline exhaust, cigarette smoke, coal tar, coal and oil combustion emissions, lubricating oils, used motor oils and crude oils and is consdered a contaminant. This compound could be used as reference material.
Catalog No. 50-256-1592