Learn More
Sigma Aldrich Fine Chemicals Biosciences Benzonase Nuclease, 9025-65-4, MFCD00131010, 25 KU
Catalog No. 501780631
Encompass
≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, storage temp: −20°C, Synonym: Endonuclease from Serratia marcescens.
Catalog No. 50-178-0631 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E101425KU
≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, storage temp: −20°C, Synonym: Endonuclease from Serratia marcescens.