Learn More
Medchemexpress LLC Benzonatate | 104-31-4 | 98.96% | 100 MG
SDP
Catalog No. 5000411620
Encompass_Preferred
Benzonatate is an orally active and non-narcotic peripheral antitussive agent. It is a reversible voltage-gated Na+ channels inhibitor and dampens the activity of cough stretch receptors.
  • Orally active
  • Non-narcotic peripheral antitussive agent
  • Reversible voltage-gated Na+ channels inhibitor
  • Dampens the activity of cough stretch receptors
Catalog No. 50-004-11620 Supplier Medchemexpress LLC Supplier No. HYB1551100MG
Benzonatate is an orally active and non-narcotic peripheral antitussive agent. It is a reversible voltage-gated Na+ channels inhibitor and dampens the activity of cough stretch receptors.
  • Orally active
  • Non-narcotic peripheral antitussive agent
  • Reversible voltage-gated Na+ channels inhibitor
  • Dampens the activity of cough stretch receptors