Learn More
Medchemexpress LLC Benzonitrile, 4,4'-(hydroxymethylene)bis- | 134521-16-7 | 234.25 | 1 ML
SDP
Catalog No. 5000445867
Encompass_Preferred
CGP 44 645 is the methanol metabolite of Letrozole. It serves as a molecular indicator for evaluating the metabolic kinetics of Letrozole in microsomes in vitro, with levels in rat liver microsomes potentially being sex-specific.
  • For research use only
  • Metabolic enzyme/protease drug metabolite
  • Molecular indicator for evaluating metabolic kinetics in microsomes in vitro
  • Soluble in DMSO (≥ 200 mg/mL)
Catalog No. 50-004-45867 Supplier Medchemexpress LLC Supplier No. HYW00337110MM/1ML
CGP 44 645 is the methanol metabolite of Letrozole. It serves as a molecular indicator for evaluating the metabolic kinetics of Letrozole in microsomes in vitro, with levels in rat liver microsomes potentially being sex-specific.
  • For research use only
  • Metabolic enzyme/protease drug metabolite
  • Molecular indicator for evaluating metabolic kinetics in microsomes in vitro
  • Soluble in DMSO (≥ 200 mg/mL)