Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Benzonitrile, 4-[(4S)-6-chloro-4-cyclopropyl-tetrahydro-2-oxo-3-(trifluoroethyl)-4-quinazolin-4-yl] | 910484-28-5 | 100.0% | 405.80 g·mol⁻¹ | C20H15ClF3N3O | 25 MG
SDP

Catalog No. 5000244130
Encompass_Preferred

TTA-Q6 is a small-molecule selective T-type calcium channel antagonist supplied for research use in neurological and electrophysiology studies. It is provided as a solid with characterization data for purity, molecular weight, and solubility to support experimental use.

  • Selective T-type Ca2+ channel antagonist for research applications.
  • High purity: 99.97% by manufacturer analysis.
  • Molecular weight 405.80 g·mol⁻¹ and formula C20H15ClF3N3O.
  • Physical form: solid, light yellow to yellow.
  • Soluble in DMSO at 125 mg/mL; examples of in vivo formulations reported.
  • Storage: powder at -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes (for example, 5 mg, 10 mg, 25 mg, 50 mg).

Catalog No. 50-002-44130 Supplier Medchemexpress LLC Supplier No. HY1038825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TTA-Q6 is a small-molecule selective T-type calcium channel antagonist supplied for research use in neurological and electrophysiology studies. It is provided as a solid with characterization data for purity, molecular weight, and solubility to support experimental use.

  • Selective T-type Ca2+ channel antagonist for research applications.
  • High purity: 99.97% by manufacturer analysis.
  • Molecular weight 405.80 g·mol⁻¹ and formula C20H15ClF3N3O.
  • Physical form: solid, light yellow to yellow.
  • Soluble in DMSO at 125 mg/mL; examples of in vivo formulations reported.
  • Storage: powder at -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes (for example, 5 mg, 10 mg, 25 mg, 50 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.