Learn More
Medchemexpress LLC Benzonitrile, 4-[5-chloro-7-[[(1R)-1,2,2-trimethylpropyl]amino]triazolo[1,5-a]pyrimidin-6-yl]-3,5-difluoro- | 2699937-76-1 | 99.5% | 390.82 | 5 MG
SDP
Catalog No. 5000387325
Encompass_Preferred
Microtubule stabilizer-1 (compound 69) is a brain-penetrant microtubule (MT) stabilizer. It increases acetylated α-tubulin (AcTub) levels and prevents the characteristic loss of neuronal MTs observed after incubation with Okadaic acid. This compound can be used for the study of Alzheimer's disease (AD) and related neurodegenerative tauopathies.
  • Brain-penetrant microtubule (MT) stabilizer
  • Increases acetylated α-tubulin (AcTub) levels
  • Prevents loss of neuronal MTs after Okadaic acid incubation
  • Used for Alzheimer's disease (AD) and neurodegenerative tauopathies research
Catalog No. 50-003-87325 Supplier Medchemexpress LLC Supplier No. HY1765405MG
Microtubule stabilizer-1 (compound 69) is a brain-penetrant microtubule (MT) stabilizer. It increases acetylated α-tubulin (AcTub) levels and prevents the characteristic loss of neuronal MTs observed after incubation with Okadaic acid. This compound can be used for the study of Alzheimer's disease (AD) and related neurodegenerative tauopathies.
  • Brain-penetrant microtubule (MT) stabilizer
  • Increases acetylated α-tubulin (AcTub) levels
  • Prevents loss of neuronal MTs after Okadaic acid incubation
  • Used for Alzheimer's disease (AD) and neurodegenerative tauopathies research