Learn More
Selleck Chemical LLC Benzophenone S4438-100mg
SDP
Catalog No. 503109491
Encompass_Preferred
Benzophenone (Diphenyl ketone Benzoylbenzene) filters out both UVA and UVB rays and is widely used in a great variety of sunscreens and personal care products
Catalog No. 50-310-9491 Supplier Selleck Chemical LLC Supplier No. S4438100MG
Benzophenone (Diphenyl ketone Benzoylbenzene) filters out both UVA and UVB rays and is widely used in a great variety of sunscreens and personal care products