Learn More
Medchemexpress LLC Benzophenone | 119-61-9 | MFCD00003076 | 182.22 | C13H10O | 500g
SDP
Catalog No. 5000269285
Encompass_Preferred
Benzophenone is an endogenous metabolite Benzophenone is a photosensitizer that absorbs UV light transfers its energy to DNA and triggers DNA damage Benzophenone can be used as the fragrance enhancer ultraviolet curing agent additive in plastics or the flavor ingredient Benzophenone can also be used in the manufacturing of insecticides agricultural chemicals hypnotics antihistamines and other pharmaceuticals Benzophenone exhibits certain carcinogenicity in mouse/rat models under long term exposure Benzophenone exhibits estrogenic activity[1][2]
Catalog No. 50-002-69285 Supplier Medchemexpress LLC Supplier No. HYY0546500G
Benzophenone is an endogenous metabolite Benzophenone is a photosensitizer that absorbs UV light transfers its energy to DNA and triggers DNA damage Benzophenone can be used as the fragrance enhancer ultraviolet curing agent additive in plastics or the flavor ingredient Benzophenone can also be used in the manufacturing of insecticides agricultural chemicals hypnotics antihistamines and other pharmaceuticals Benzophenone exhibits certain carcinogenicity in mouse/rat models under long term exposure Benzophenone exhibits estrogenic activity[1][2]